Learn More
Thomas Scientific Centrifuge Tube Stand
SDP
Catalog No. 501460915
Encompass_Preferred
Foxx Life Sciences' Centrifuge Tube Stands are intended for use with centrifuge tubes. They are made from Class VI, FDA Grade, BPA-Free Polystyrene, and are conveniently sold in packs of 5. Autofil tube stand can hold one 15ml or 50ml tube Can also be used with the Autofil Centrifuge Filtration system
Catalog No. 50-146-0915 Supplier Thomas Scientific Supplier No. 1171Z58
Foxx Life Sciences' Centrifuge Tube Stands are intended for use with centrifuge tubes. They are made from Class VI, FDA Grade, BPA-Free Polystyrene, and are conveniently sold in packs of 5. Autofil tube stand can hold one 15ml or 50ml tube Can also be used with the Autofil Centrifuge Filtration system