Learn More
Mascon Medical Mascon Medical™ Centrifuge Tube With Looped Flat Cap, 1.5ml, 500EA/Bag, 10 Bags/Case
SDP
Catalog No. NC2458303
Encompass_Preferred
Mascon Medical™ 1.5mL high-performance micro Centrifuge Tubes are made of high-quality, medical-grade polypropylene which can withstand autoclaving, boiling, freezing, and high-speed centrifugation. Microcentrifuge tubes can be used with various brands of centrifuges and can be widely applied in research in such fields as medical examination, molecular biology, and genetic biology. Microcentrifuge tubes can be used with various brands of centrifuges and can be widely applied in research for processes such as medical examination, molecular biology, and genetic biology.Features:•Looped flat cap•Meets ISO13485 quality management system standard•Conical bottom•DNase/RNase free•Non-pyrogenic•Frosted area for marking capabilities•Chemical corrosion resistantSpecifications:•Volume: 1.5mL•Qty. / Case: 5,000•Color: Transparent•Material: Polypropylene•Operating Temperature: -80℃~121℃•Maximum RCF: 30,000g•Size: Φ10.5mm*45mm•Packaging Format: 500EA/Bag, 10Bags/Case
Catalog No. NC2458303 Supplier Mascon Medical Supplier No. FSCFTUBE1.55000CS
Mascon Medical™ 1.5mL high-performance micro Centrifuge Tubes are made of high-quality, medical-grade polypropylene which can withstand autoclaving, boiling, freezing, and high-speed centrifugation. Microcentrifuge tubes can be used with various brands of centrifuges and can be widely applied in research in such fields as medical examination, molecular biology, and genetic biology. Microcentrifuge tubes can be used with various brands of centrifuges and can be widely applied in research for processes such as medical examination, molecular biology, and genetic biology.Features:•Looped flat cap•Meets ISO13485 quality management system standard•Conical bottom•DNase/RNase free•Non-pyrogenic•Frosted area for marking capabilities•Chemical corrosion resistantSpecifications:•Volume: 1.5mL•Qty. / Case: 5,000•Color: Transparent•Material: Polypropylene•Operating Temperature: -80℃~121℃•Maximum RCF: 30,000g•Size: Φ10.5mm*45mm•Packaging Format: 500EA/Bag, 10Bags/Case