Learn More
Mascon Medical Centrifuge Tube, Conical Bottom, Flat Cap, Polypropylene, 15ml, 50EA/Bag, 10Bags/Case
SDP
Catalog No. NC2180275
Encompass_Preferred
Mascon Medical™ Conical Bottom Centrifuge Tubes are engineered to provide a high strength product with vast temperature stability, ensuring outstanding performance in your critical applications. These 15ml Conical Bottom Tubes with flat caps are ideal for cell centrifugation, pelleting, separation by density gradients. These leak proof centrifuge tubes are made of ultra-clear polypropylene, have white printed graduations with a large white marking strip for labeling and are Non-Pyrogenic and DNase/RNase free.Specifications: Volume: 15ml Qty. / Case: 500 Material: Polypropylene Cap Material: HDPE Color: Clear with Blue Cap Packaging Format: 50EA/Bag, 10Bags/CaseFeatures: Non-Pyrogenic DNase/RNase free Conical Bottom Flat Cap Leak proof
This item is not returnable. View return policy
Catalog No. NC2180275 Supplier Mascon Medical Supplier No. FSCFTUBE15
Mascon Medical™ Conical Bottom Centrifuge Tubes are engineered to provide a high strength product with vast temperature stability, ensuring outstanding performance in your critical applications. These 15ml Conical Bottom Tubes with flat caps are ideal for cell centrifugation, pelleting, separation by density gradients. These leak proof centrifuge tubes are made of ultra-clear polypropylene, have white printed graduations with a large white marking strip for labeling and are Non-Pyrogenic and DNase/RNase free.Specifications: Volume: 15ml Qty. / Case: 500 Material: Polypropylene Cap Material: HDPE Color: Clear with Blue Cap Packaging Format: 50EA/Bag, 10Bags/CaseFeatures: Non-Pyrogenic DNase/RNase free Conical Bottom Flat Cap Leak proof