Learn More
Electron Microscopy Sciences Flat Screw Cap Conical Centrifuge Tube, 15 ml, Sterile Bag, 17 x 118 mm
SDP
Catalog No. 501929173
Encompass_Preferred
Centrifuge tubes made of premium polypropylene for clarity and centrifugal strength. They are supplied sterile, and although they are autoclavable, their polyethylene screw caps are not. Tubes and caps may be frozen at temperatures as low as -45 deg C. Caps have a flat top surface for easy marking. Sterile, Rnase, Dnase, pyrogen, and cytotoxin free. Compatible with standard rotors. Frosted marking surface. Maximum centrifugal force is 12,000xg.
Volume: 15 ml
Package: Sterile Bag
Size: 17 x 118 mm
Catalog No. 50-192-9173 Supplier Electron Microscopy Sciences Supplier No. 6302602
Centrifuge tubes made of premium polypropylene for clarity and centrifugal strength. They are supplied sterile, and although they are autoclavable, their polyethylene screw caps are not. Tubes and caps may be frozen at temperatures as low as -45 deg C. Caps have a flat top surface for easy marking. Sterile, Rnase, Dnase, pyrogen, and cytotoxin free. Compatible with standard rotors. Frosted marking surface. Maximum centrifugal force is 12,000xg.
Volume: 15 ml
Package: Sterile Bag
Size: 17 x 118 mm