Learn More
Chemglass Life Sciences Centrifuge Tube, 15mL, Graduated, Polypropylene, Sterile
SDP
Catalog No. 501897628
Encompass
• Printed graduations (with marking area)• Polypropylene for the tube and polyethylene for the cap• Flat markable cap with leak-proof seal• One-hand operation design for the caps provides easy handling• Sterilized by E-Beam• Non-pyrogenic, DNase- and RNase-free• Packed in bags of 50
Catalog No. 50-189-7628 Supplier Chemglass Life Sciences Supplier No. CGE4303015
• Printed graduations (with marking area)• Polypropylene for the tube and polyethylene for the cap• Flat markable cap with leak-proof seal• One-hand operation design for the caps provides easy handling• Sterilized by E-Beam• Non-pyrogenic, DNase- and RNase-free• Packed in bags of 50