Learn More
Chemglass Life Sciences Chemglass 15 mL Graduated Centrifuge Tube, Polypropylene, Flat Markable Cap, Non-sterile, Bulk Pack
SDP
Catalog No. 502733410
Encompass_Preferred
Chemglass 15 mL centrifuge tubes are designed for routine laboratory centrifugation, sample storage, and processing. These tubes feature printed graduations and a flat, markable cap for easy identification and handling. They are constructed from polypropylene with polyethylene caps and are supplied non-sterile in bulk packs.
  • 15 mL capacity with printed graduations and marking area.
  • Polypropylene tube; polyethylene cap.
  • Non-sterile, suitable for general laboratory applications.
  • Flat markable cap with leak-proof seal and one-hand operation design.
  • Bulk packaging, packed in bags of 50; tubes and caps supplied separately.
Catalog No. 50-273-3410 Supplier Chemglass Life Sciences Supplier No. CGN4303115
Chemglass 15 mL centrifuge tubes are designed for routine laboratory centrifugation, sample storage, and processing. These tubes feature printed graduations and a flat, markable cap for easy identification and handling. They are constructed from polypropylene with polyethylene caps and are supplied non-sterile in bulk packs.
  • 15 mL capacity with printed graduations and marking area.
  • Polypropylene tube; polyethylene cap.
  • Non-sterile, suitable for general laboratory applications.
  • Flat markable cap with leak-proof seal and one-hand operation design.
  • Bulk packaging, packed in bags of 50; tubes and caps supplied separately.