Learn More
Cole-Parmer CENTRIFUGE TUBE 15ML VENT CAP
Supplier:  Cole-Parmer 0633640
Encompass_Preferred
Convenient and economical option for small scale, high density suspension cell cultureFeature a vented cap with a 0.22 μm hydrophobic membrane—ideal for biological reaction experiments that require aseptic conditions with free gas exchangeBlack graduations make reading liquid-levels easyLarge, white marking area simplifies labeling
Catalog No. NC2547326