Learn More
Chemglass Life Sciences Tube, Centrifuge, 50mL, Conical Bottom, 24-415 GPI Thread, 28mm OD x 155mm Length
SDP
Catalog No. CG401301
Encompass_Preferred
Tube, Centrifuge, 50mL, Conical Bottom, 24-415 GPI Thread, 28mm OD x 155mm Length. Centrifuge tube with a short, conical bottom and 24-415 GPI threaded top. Fits I.E.C. shield number 320. Supplied complete with a Poly-Seal polypropylene lined phenolic screw cap.
Catalog No. CG401301 Supplier Chemglass Life Sciences Supplier No. CG401301
Tube, Centrifuge, 50mL, Conical Bottom, 24-415 GPI Thread, 28mm OD x 155mm Length. Centrifuge tube with a short, conical bottom and 24-415 GPI threaded top. Fits I.E.C. shield number 320. Supplied complete with a Poly-Seal polypropylene lined phenolic screw cap.