Learn More
Electron Microscopy Sciences Centrifuge tubes made of polypropylene for clarity and centrifugal strength. Tubes autoclavable but screw caps are not. Sterile, RNase, DNase, pyrogen, and cytotoxin free. Frosted marking surface.
SDP
Catalog No. 501929174
Encompass_Preferred
Centrifuge tubes made of premium polypropylene for clarity and centrifugal strength. They are supplied sterile, and although they are autoclavable, their polyethylene screw caps are not. Tubes and caps may be frozen at temperatures as low as -45 deg C. Caps have a flat top surface for easy marking. Sterile, Rnase, Dnase, pyrogen, and cytotoxin free. Compatible with standard rotors. Frosted marking surface.
Catalog No. 50-192-9174 Supplier Electron Microscopy Sciences Supplier No. 6302603
Centrifuge tubes made of premium polypropylene for clarity and centrifugal strength. They are supplied sterile, and although they are autoclavable, their polyethylene screw caps are not. Tubes and caps may be frozen at temperatures as low as -45 deg C. Caps have a flat top surface for easy marking. Sterile, Rnase, Dnase, pyrogen, and cytotoxin free. Compatible with standard rotors. Frosted marking surface.