Learn More
Electron Microscopy Sciences Flat Screw Cap Conical Centrifuge Tube, 50 ml, Sterile Bag, 29x 115 mm
SDP
Catalog No. 501929175
Encompass_Preferred
Centrifuge tubes made of premium polypropylene for clarity and centrifugal strength. They are supplied sterile, and although they are autoclavable, their polyethylene screw caps are not. Tubes and caps may be frozen at temperatures as low as -45 deg C. Caps have a flat top surface for easy marking. Sterile, Rnase, Dnase, pyrogen, and cytotoxin free. Compatible with standard rotors. Frosted marking surface. Maximum centrifugal force is 15,000xg.
Volume: 50 ml
Package: Sterile Bag
Size: 29x 115 mm
Catalog No. 50-192-9175 Supplier Electron Microscopy Sciences Supplier No. 6302604
Centrifuge tubes made of premium polypropylene for clarity and centrifugal strength. They are supplied sterile, and although they are autoclavable, their polyethylene screw caps are not. Tubes and caps may be frozen at temperatures as low as -45 deg C. Caps have a flat top surface for easy marking. Sterile, Rnase, Dnase, pyrogen, and cytotoxin free. Compatible with standard rotors. Frosted marking surface. Maximum centrifugal force is 15,000xg.
Volume: 50 ml
Package: Sterile Bag
Size: 29x 115 mm