Learn More
Electron Microscopy Sciences Flat Screw Cap Free Standing, 50 ml, Sterile Bag, Sterile Bag package
SDP
Catalog No. 501929177
Encompass_Preferred
Centrifuge tubes made of premium polypropylene for clarity and centrifugal strength. They are supplied sterile, and although they are autoclavable, their polyethylene screw caps are not. Tubes and caps may be frozen at temperatures as low as -45 deg C. Caps have a flat top surface for easy marking. Sterile, Rnase, Dnase, pyrogen, and cytotoxin free. Compatible with standard rotors. Frosted marking surface. Skirted bottom so tube stands upright without a rack.
Volume: 50 ml
Package: Sterile Bag
Catalog No. 50-192-9177 Supplier Electron Microscopy Sciences Supplier No. 6302606
Centrifuge tubes made of premium polypropylene for clarity and centrifugal strength. They are supplied sterile, and although they are autoclavable, their polyethylene screw caps are not. Tubes and caps may be frozen at temperatures as low as -45 deg C. Caps have a flat top surface for easy marking. Sterile, Rnase, Dnase, pyrogen, and cytotoxin free. Compatible with standard rotors. Frosted marking surface. Skirted bottom so tube stands upright without a rack.
Volume: 50 ml
Package: Sterile Bag