Learn More
Thomas Scientific 50mL Centrifuge Tube, Flip-Top Cap - Bag, Sterile
SDP
Catalog No. 501459079
Encompass
CELLTREAT Flip Top centrifuge tubes are manufactured in a class 100,000 clean room and made of high-clarity polypropylene. Graduations are printed in a bold easy-to-read black and are accurate to Ý 2%. Large white marking area provides excellent contrast for identification. The ergonomic leak-free flip top cap design allows easy one hand opening of caps while preventing loss of a separate cap. Flip top caps are smooth, flat surface polyethylene. Temperature range is -80ÝC to 120ÝC. CELLTREAT 15ml and 50ml Flip Top Centrifuge tubes are available in either paperboard recyclable racks or bulk packaged in sterile re-sealable (zip closure) bags. Gamma irradiation sterilized, RNase-free, DNase-free, DNA-free, and non-pyrogenic.
This item is not returnable. View return policy
Catalog No. 50-145-9079 Supplier Thomas Scientific Supplier No. 1222B22
CELLTREAT Flip Top centrifuge tubes are manufactured in a class 100,000 clean room and made of high-clarity polypropylene. Graduations are printed in a bold easy-to-read black and are accurate to Ý 2%. Large white marking area provides excellent contrast for identification. The ergonomic leak-free flip top cap design allows easy one hand opening of caps while preventing loss of a separate cap. Flip top caps are smooth, flat surface polyethylene. Temperature range is -80ÝC to 120ÝC. CELLTREAT 15ml and 50ml Flip Top Centrifuge tubes are available in either paperboard recyclable racks or bulk packaged in sterile re-sealable (zip closure) bags. Gamma irradiation sterilized, RNase-free, DNase-free, DNA-free, and non-pyrogenic.