Learn More
Mascon Medical Centrifuge Tube, Conical Bottom, Flat Cap, 5ml, 300EA/Bag, 10Bags/Case
SDP
Catalog No. NC2180296
Encompass_Preferred
Mascon Medical™ Conical Bottom Centrifuge Tubes are engineered to provide a high strength product with vast temperature stability, ensuring outstanding performance in your critical applications. These 5ml Conical Bottom Tubes with flat caps are ideal for cell centrifugation, pelleting, separation by density gradients, and maintain good stability in the temperature range of -80C to 121C. The tube cover can be opened and closed with one hand making it especially easy to operate while providing a frosted writing area for easy identification. Non-Pyrogenic and DNase/RNase free.Specifications: Volume: 5ml Qty. / Case: 3,000 Color: Clear Packaging Format: 300EA/Bag, 10Bags/CaseFeatures: Non-Pyrogenic DNase/RNase free Conical Bottom Flat Cap Maximum Centrifugal Force, 4000 RCF Temperature Stability Range from -80C to 121C
This item is not returnable. View return policy
Catalog No. NC2180296 Supplier Mascon Medical Supplier No. FSCFTUBE5
Mascon Medical™ Conical Bottom Centrifuge Tubes are engineered to provide a high strength product with vast temperature stability, ensuring outstanding performance in your critical applications. These 5ml Conical Bottom Tubes with flat caps are ideal for cell centrifugation, pelleting, separation by density gradients, and maintain good stability in the temperature range of -80C to 121C. The tube cover can be opened and closed with one hand making it especially easy to operate while providing a frosted writing area for easy identification. Non-Pyrogenic and DNase/RNase free.Specifications: Volume: 5ml Qty. / Case: 3,000 Color: Clear Packaging Format: 300EA/Bag, 10Bags/CaseFeatures: Non-Pyrogenic DNase/RNase free Conical Bottom Flat Cap Maximum Centrifugal Force, 4000 RCF Temperature Stability Range from -80C to 121C