Learn More
MTC Bio Snap Cap 5mL MacroTubes, sterile, ten bags of 20 tubes (200 total)
Catalog No. NC1299551
Encompass_Preferred
This item has a minimum qty of 4 per supplier requirements.
MTC Bios new 5mL Five-O™ MacroTubes™ take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption. Useful from -86C to 110C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from gamma sterilized racked packaging with attached caps or non-sterile bulk packaging with separate caps. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space.
Catalog No. NC1299551 Supplier MTC Bio Supplier No. C2520
This item has a minimum qty of 4 per supplier requirements.
MTC Bios new 5mL Five-O™ MacroTubes™ take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption. Useful from -86C to 110C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from gamma sterilized racked packaging with attached caps or non-sterile bulk packaging with separate caps. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space.