Learn More
Electron Microscopy Sciences SpectraTube™ Centrifuge Tubes 50 ml, 29 x 115 mm
SDP
Catalog No. 501929300
Encompass_Preferred
The SpectraTube centrifuge tubes have color coded caps for easy sample identification and are perfect for any lab conducting multiple studies, day of the week studies, or any experiments requiring multiple sample types or solutions. All caps are flat, brightly colored, and frosted to make them easy to write on. Made of premium polyethylene, with exceptionally uniform wall thickness for clarity and centrifugal strength. Tubes and caps may be used for sample storage at temperaturess as low as -80 deg C. Tubes and caps are suitable for stem cell research and certified sterile, RNase, Dnase, pyrogen and cytotoxin free. Sterile (gamma) and individually packed. Leak proof. Maximum centrifugal force is 15,000 xg.
Size: 29 x 115 mm
Catalog No. 50-192-9300 Supplier Electron Microscopy Sciences Supplier No. 6302803
The SpectraTube centrifuge tubes have color coded caps for easy sample identification and are perfect for any lab conducting multiple studies, day of the week studies, or any experiments requiring multiple sample types or solutions. All caps are flat, brightly colored, and frosted to make them easy to write on. Made of premium polyethylene, with exceptionally uniform wall thickness for clarity and centrifugal strength. Tubes and caps may be used for sample storage at temperaturess as low as -80 deg C. Tubes and caps are suitable for stem cell research and certified sterile, RNase, Dnase, pyrogen and cytotoxin free. Sterile (gamma) and individually packed. Leak proof. Maximum centrifugal force is 15,000 xg.
Size: 29 x 115 mm