Learn More
Mascon Medical Mascon Medical™ Centrifuge Tube Rack, 15ml-50ml, Orange, 50EA/Case
SDP
Catalog No. NC2458305
Encompass_Preferred
Mascon Medical™ Centrifuge Tube Racks provide high-performance storage of 15ml and 50ml centrifugal tubes. Features:•Meets ISO13485 quality management system standardSpecifications:•Volume: 24-well 15ml, 30-well 50ml•Qty. / Case: 50•Color: Orange•Material: Polypropylene•Temperature Range: -40℃~100℃•Size: 207*172*60mm•Packaging Format: 1EA/Bag, 50Bags/Case
Catalog No. NC2458305 Supplier Mascon Medical Supplier No. FSCFTUBERACK
Mascon Medical™ Centrifuge Tube Racks provide high-performance storage of 15ml and 50ml centrifugal tubes. Features:•Meets ISO13485 quality management system standardSpecifications:•Volume: 24-well 15ml, 30-well 50ml•Qty. / Case: 50•Color: Orange•Material: Polypropylene•Temperature Range: -40℃~100℃•Size: 207*172*60mm•Packaging Format: 1EA/Bag, 50Bags/Case