Learn More
Research Products International Corp Poxygrid Centrifuge Tube Rack, 8 Places
SDP
Catalog No. 50212939
Encompass
Durable epoxy coated steel wire racks are designed to hold 50 ml centrifuge tubes. Heat-cured epoxy coating is unaffected by organic solvents, salts, weak acids and weak alkalis. Racks feature dual tiers and rail channels for maximum tube stability.
WARNING  This product can expose you to chemicals including Titanium Dioxide, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-212-939 Supplier Research Products International Corp Supplier No. 247940
Durable epoxy coated steel wire racks are designed to hold 50 ml centrifuge tubes. Heat-cured epoxy coating is unaffected by organic solvents, salts, weak acids and weak alkalis. Racks feature dual tiers and rail channels for maximum tube stability.
WARNING  This product can expose you to chemicals including Titanium Dioxide, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.