Learn More
Bellco Glass, Inc. Centrifuge Tube Stand, 15ml and 50 ml, 5/PK
SDP
Catalog No. 502550296
Encompass_Preferred
Disposability Saves Time and Lab Funds' Centrifuge Tube Stands are intended for use with sold separately Autofil centrifuge tubes. Made from Class VI, FDA Grade, BPA-Free Polystyrene, in economical packs of 5, these disposable centrifuge tube stands not only save you valuable time, but also the expense of autoclaving. Autofil tube stand can hold one 15ml or 50ml tube.Can also be used with the Autofil Centrifuge Filtration system.Autofil Disposable Centrifuge Tube Stand - 5 Pack Autofil Disposable Centrifuge Tube Stand - 5 Pack Autofil Disposable Centrifuge Tube Stand - 5 PackSpecificationsColor: WhiteQuantity: 5/pk Material: Polystyrene
Catalog No. 50-255-0296 Supplier Bellco Glass, Inc. Supplier No. 452800466
Disposability Saves Time and Lab Funds' Centrifuge Tube Stands are intended for use with sold separately Autofil centrifuge tubes. Made from Class VI, FDA Grade, BPA-Free Polystyrene, in economical packs of 5, these disposable centrifuge tube stands not only save you valuable time, but also the expense of autoclaving. Autofil tube stand can hold one 15ml or 50ml tube.Can also be used with the Autofil Centrifuge Filtration system.Autofil Disposable Centrifuge Tube Stand - 5 Pack Autofil Disposable Centrifuge Tube Stand - 5 Pack Autofil Disposable Centrifuge Tube Stand - 5 PackSpecificationsColor: WhiteQuantity: 5/pk Material: Polystyrene