Learn More
CELLTREAT Scientific Products Reusable Multi-purpose Centrifuge Tube Stand, Non-sterile
SDP
Catalog No. 50607512
Encompass_Preferred
CELLTREAT Scientific Products Multi-purpose Centrifuge Tube Stand. Multi-purpose Centrifuge Tube Stand securely holds tube during filtration and includes molded positions for additional tube sizes. Centrifuge Tube Stand will hold one 50mL tube, three 15mL tubes and three microcentrifuge tubes (1.5mL). Centrifuge Tube Stands are manufactured from polypropylene and are autoclavable, reusable and recyclable. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/bag, 5/case.
Catalog No. 50-607-512 Supplier CELLTREAT Scientific Products Supplier No. 229409
CELLTREAT Scientific Products Multi-purpose Centrifuge Tube Stand. Multi-purpose Centrifuge Tube Stand securely holds tube during filtration and includes molded positions for additional tube sizes. Centrifuge Tube Stand will hold one 50mL tube, three 15mL tubes and three microcentrifuge tubes (1.5mL). Centrifuge Tube Stands are manufactured from polypropylene and are autoclavable, reusable and recyclable. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/bag, 5/case.