Learn More
Alkali Scientific ASI™ Centrifuge Tubes, 25mL, 8 foam racks of 25 tubes, 200 per pack
SDP
Catalog No. 503027053
Encompass_Preferred
ASI™ Centrifuge Tubes   Screw-Cap polypropylene tubes are manufactured for temperature and chemical resistance. These tubes are intended for large volume centrifugation and are widely used in cell harvesting/purification and drug discovery applications. They are supplied in sterile, RNase, DNase, and pyrogen-free packs of 50 tubes per sterile bag, 1000 per case. Sterile, RNase, DNase, pyrogen and cytotoxin free Available racked or bagged Compatible with standard rotors Frosted marking surface
Catalog No. 50-302-7053 Supplier Alkali Scientific Supplier No. DP450
ASI™ Centrifuge Tubes   Screw-Cap polypropylene tubes are manufactured for temperature and chemical resistance. These tubes are intended for large volume centrifugation and are widely used in cell harvesting/purification and drug discovery applications. They are supplied in sterile, RNase, DNase, and pyrogen-free packs of 50 tubes per sterile bag, 1000 per case. Sterile, RNase, DNase, pyrogen and cytotoxin free Available racked or bagged Compatible with standard rotors Frosted marking surface