Learn More
CELLTREAT Scientific Products PLASTIC RACK ONLY, 15mL Centrifuge Tube, Best Value RackMaster, Non-sterile, Reusable
SDP
Catalog No. NC3726757
Encompass_Preferred
RackMaster plastic carrier protects paperboard centrifuge tube racks from deterioration caused by wet lab surfaces
Bonus side compartments offer space to hold markers, caps, and additional tubes
Refill RackMasters with paperboard centrifuge tube racks to reduce waste and save money compared to foam racks
Plastic racks are reusable and autoclavable; paperboard racks are recyclable
Available in 15mL and 50mL configurations
Catalog No. NC3726757 Supplier CELLTREAT Scientific Products Supplier No. 230469
RackMaster plastic carrier protects paperboard centrifuge tube racks from deterioration caused by wet lab surfaces
Bonus side compartments offer space to hold markers, caps, and additional tubes
Refill RackMasters with paperboard centrifuge tube racks to reduce waste and save money compared to foam racks
Plastic racks are reusable and autoclavable; paperboard racks are recyclable
Available in 15mL and 50mL configurations