Learn More
Beckman Coulter Open-Top Thickwall Polycarbonate Open-Top Tube, 50 mL, 29 x 104mm, 25/pk
Catalog No. NC9021042
Encompass_Preferred
Polycarbonate Open-Top Tube For use in High Performance and High Capacity Rotors: JA-30.50 Ti; At 2C; at 25C, maximum speed must be limited to 29 000 rpm for tube.
Catalog No. NC9021042 Supplier Beckman Coulter Supplier No. 363647
Polycarbonate Open-Top Tube For use in High Performance and High Capacity Rotors: JA-30.50 Ti; At 2C; at 25C, maximum speed must be limited to 29 000 rpm for tube.