Learn More
Alkali Scientific ASI™ Centrifuge Tubes, 50mL, non-sterile bulk bag, 500 per pack
SDP
Catalog No. 503026457
Encompass_Preferred
ASI™ Centrifuge Tubes   Screw-Cap polypropylene tubes are manufactured for temperature and chemical resistance. These tubes are intended for large volume centrifugation and are widely used in cell harvesting/purification and drug discovery applications. They are supplied in sterile, RNase, DNase, and pyrogen-free packs of 50 tubes per sterile bag, 1000 per case. Sterile, RNase, DNase, pyrogen and cytotoxin free Available racked or bagged Compatible with standard rotors Frosted marking surface
Catalog No. 50-302-6457 Supplier Alkali Scientific Supplier No. C5608
ASI™ Centrifuge Tubes   Screw-Cap polypropylene tubes are manufactured for temperature and chemical resistance. These tubes are intended for large volume centrifugation and are widely used in cell harvesting/purification and drug discovery applications. They are supplied in sterile, RNase, DNase, and pyrogen-free packs of 50 tubes per sterile bag, 1000 per case. Sterile, RNase, DNase, pyrogen and cytotoxin free Available racked or bagged Compatible with standard rotors Frosted marking surface