Learn More
TWD Scientific LLC Tube, Centrifuge, Twdbiox, 50Ml, Sterile, Printed Graduations, Wrinting Patch, Polypropylene, Plug-Seal Screw Assembled, Conical Bottom, 500 Tubes/Case.
SDP
Catalog No. 502285025 Shop All TWD Scientific LLC Products
Encompass_Preferred
50Ml Clear Polypropylene Sterile Conical Bottom Centrifuge Tubes With Plug-Seal Screw Caps Assembled. Tubes Have Writing Patch And Marked Graduations. 25 Tubes/Pack, 20 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5025 Supplier TWD Scientific LLC Supplier No. BX100502
50Ml Clear Polypropylene Sterile Conical Bottom Centrifuge Tubes With Plug-Seal Screw Caps Assembled. Tubes Have Writing Patch And Marked Graduations. 25 Tubes/Pack, 20 Packs/Case, Price/Case