Learn More
TWD Scientific LLC Tube, Centrifuge, Twdbiox, 50Ml, Sterile, Printed Graduations, Wrinting Patch, Polypropylene, Plug-Seal Screw Assembled, Conical Bottom, Tubes Loaded In Rack, 500 Tubes/Case.
SDP
Catalog No. 502285024 Shop All TWD Scientific LLC Products
Encompass_Preferred
50Ml Clear Polypropylene Sterile Conical Bottom Centrifuge Tubes With Plug-Seal Screw Caps Assembled. Tubes Have Writing Patch And Marked Graduations. Tubes Come Loaded Into Rack. 25 Tubes/Rack, 20 Racks/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5024 Supplier TWD Scientific LLC Supplier No. BX100501
50Ml Clear Polypropylene Sterile Conical Bottom Centrifuge Tubes With Plug-Seal Screw Caps Assembled. Tubes Have Writing Patch And Marked Graduations. Tubes Come Loaded Into Rack. 25 Tubes/Rack, 20 Racks/Case, Price/Case