Learn More
Beckman Coulter CENTRIFUGE TUBES PC
Catalog No. NC9867320
Encompass_Preferred
Thickwall Polycarbonate Tubes,Size:29 x 104, Vol.(mL): 40.0
Catalog No. NC9867320 Supplier Beckman Coulter Supplier No. 363664
Thickwall Polycarbonate Tubes,Size:29 x 104, Vol.(mL): 40.0