Learn More
MTC Bio Clinical Centrifuge with 6x10ml swing out rotor, accepts all common blood tubes up to 10ml. Designed for blood, urine and cell separations.
Catalog No. NC1827679
Encompass_Preferred
Centrifuge, Clinical Centrifuge with 6x10ml swing-out rotor, accepts common blood tubes up to 10ml, 99 minute timer, digital display, 3400rpm/1600xg, UL listed, designed for blood, urine and cell separations.
Catalog No. NC1827679 Supplier MTC Bio Supplier No. TB50006H
Centrifuge, Clinical Centrifuge with 6x10ml swing-out rotor, accepts common blood tubes up to 10ml, 99 minute timer, digital display, 3400rpm/1600xg, UL listed, designed for blood, urine and cell separations.