Learn More
GRANT INSTRUMENTS Centrifuge/vortex mixer 300-1,500rpm, vortex 300-1,200rpm rotor to hold 2 PCR plates
SDP
Catalog No. NC1820421
Encompass_Preferred
CVP-2 is an all-in-one PCR plate centrifuge / vortex mixer that allows for the simultaneous sample preparation of multiple samples at one time. CVP-2 USA - a unit with US plug.Versatile through being able to hold un, semi and fully skirted PCR plates with no additional accessories required.The CVP-2 offers 3 devices in one:Centrifuge with vortex mixingPCR plate centrifugePCR plate mixerCVP-2 – Key features:Centrifuge and vortex mixer combined for significant time savingCentrifugation mixing speed: 300 to 1500 rpmIndependent vortex and centrifuge timers with up to 999 cyclesAdjustable rpm or 4 programmable presetsConsistently prepare up to 192 samples continuously
Catalog No. NC1820421 Supplier GRANT INSTRUMENTS Supplier No. CVP2
CVP-2 is an all-in-one PCR plate centrifuge / vortex mixer that allows for the simultaneous sample preparation of multiple samples at one time. CVP-2 USA - a unit with US plug.Versatile through being able to hold un, semi and fully skirted PCR plates with no additional accessories required.The CVP-2 offers 3 devices in one:Centrifuge with vortex mixingPCR plate centrifugePCR plate mixerCVP-2 – Key features:Centrifuge and vortex mixer combined for significant time savingCentrifugation mixing speed: 300 to 1500 rpmIndependent vortex and centrifuge timers with up to 999 cyclesAdjustable rpm or 4 programmable presetsConsistently prepare up to 192 samples continuously