Learn More
Grainger Centrifuge with Rotor Benchtop
Catalog No. 502750464
Encompass_Preferred
The Benchmark Scientific Mini Centrifuge C1012 is a compact benchtop centrifuge designed for quick spin downs and separations in tubes up to 15mL. It has a maximum speed of 6,000rpm and maximum RCF of 2,000 x g. The centrifuge has a built-in 30-minute mechanical timer and accommodates 2 x 15mL tubes. It features a brushless motor for quiet operation and has a lightweight, compact footprint measuring 8.5"W x 11"D x 5.75"H. Safety features include an emergency lid lock and imbalance detection that prevents the centrifuge from operating if improperly loaded.
Catalog No. 50-275-0464 Supplier Grainger Supplier No. 14N155
The Benchmark Scientific Mini Centrifuge C1012 is a compact benchtop centrifuge designed for quick spin downs and separations in tubes up to 15mL. It has a maximum speed of 6,000rpm and maximum RCF of 2,000 x g. The centrifuge has a built-in 30-minute mechanical timer and accommodates 2 x 15mL tubes. It features a brushless motor for quiet operation and has a lightweight, compact footprint measuring 8.5"W x 11"D x 5.75"H. Safety features include an emergency lid lock and imbalance detection that prevents the centrifuge from operating if improperly loaded.