Learn More
Medchemexpress LLC HY-18683 10mg Medchemexpress, Centrinone-B CAS:1798871-31-4 Purity:>98%
SDP
Catalog No. 502029264
Encompass_Preferred
Medchemexpress, HY-18683 10mg Centrinone-B CAS:1798871-31-4 Centrinone-B (LCR-323) is a potent and highly selective PLK4 inhibitor, with a Ki of 0.59 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9264 Supplier Medchemexpress LLC Supplier No. HY1868310MG
Medchemexpress, HY-18683 10mg Centrinone-B CAS:1798871-31-4 Centrinone-B (LCR-323) is a potent and highly selective PLK4 inhibitor, with a Ki of 0.59 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.