Learn More
Selleck Chemical LLC Centrinone LCR-263-5mg
SDP
Supplier:  Selleck Chemical LLC S78375MG
Encompass_Preferred
Centrinone (LCR-263) is a selective and reversible polo-like kinase 4 (PLK4) inhibitor with Ki of 0.16 nM.
Catalog No. 50-239-0064