Learn More
Thermo Scientific™ CentriPAK™ Accessory for Sorvall BIOS 16 Bioprocessing Centrifuge
Shop All Thermo Scientific ProductsDescription
Accessory for Thermo Scientific™ CentriPAK™ BioProcess Container (BPC). CentriPAK BPC is designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor.
Thermo Scientific™ CentriPAK™ BioProcess Container (BPC) enables you to:
- Reduce cross-contamination risk while separating cell cultures
- Perform gentle, high-throughput and high-efficiency harvesting through centrifugation
- Eliminate post-use cleaning steps required with reusable containers
- Meet rigorous quality control and assurance practices for the manufacture of industry-standard material components and aseptic product contact film
Specifications
Specifications
| Type | CentriPAK Accessory |
| Product Line | Polaris |
| Material | Stainless Steel |
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | CentriPAK BioProcess Container |
| Description | CentriPAK BPC Hanging Rack |
| No. per Pack | 1 Ea. |