Learn More
Description
Includes
Rectangular Frame, Support Legs (2), Rotating Assembly consisting of Rotational Shaft, Encoder Wheel, 3-Step Pulley, Rotating Beam, Bearing Housing, Sliding Carriage to hold the test masses, Counterbalance Carriage to balance the rotating beam, Swivel Assembly (connects Sliding Carriage to a Dual-Range Force Sensor), Ultra Pulley, Ultra Pulley Bracket (preinstalled on Assembly), Photogate Bracket with 1/4 x 20 mounting screw (preinstalled on Assembly), Dual-Range Force Sensor Bracket (preinstalled on Assembly), 50 g masses (4), 100 g masses (4), Hook (attaches a Wireless Dynamic Sensor System to the Sliding Carriage), WDSS Mounting Screw and T Nut to secure a WDSS
- A force sensor measures the centripetal force exerted on a mass as it moves in a circle. A photogate can be used to measure the angular speed. Different masses can be added to the platform and positioned at variable radii. The platform can be driven by a falling mass. The apparatus also accommodates the Wireless Dynamic Sensor System to measure force, which provides the best set of data. Sensors are sold separately.
- Includes:
- Rectangular Frame
- Support Legs (2)
- Rotating Assembly consisting of Rotational Shaft, Encoder Wheel, 3-Step Pulley, Rotating Beam, Bearing Housing
- Sliding Carriage to hold the test masses
- Counterbalance Carriage to balance the rotating beam
- Swivel Assembly (connects Sliding Carriage to a Dual-Range Force Sensor)
- Ultra Pulley
- Ultra Pulley Bracket (preinstalled on Assembly)
- Photogate Bracket with 1/4 x 20 mounting screw (preinstalled on Assembly)
- Dual-Range Force Sensor Bracket (preinstalled on Assembly)
- 50 g masses (4)
- 100 g masses (4)
- Hook (attaches a Wireless Dynamic Sensor System to the Sliding Carriage)
- WDSS Mounting Screw and T Nut to secure a WDSS
Warranty and Services
5-year limited warranty
Specifications
Specifications
| Grade | High School and College |
| For Use With (Application) | Educational use only |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

