Learn More
Brady Centurion™ Flood Bags
Catalog No. 17033785 Shop All Brady Corporation Products
Change view
Click to view available options
Packaging Quantity:
100/Cs.
200/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
17-033-785 200/Cs.
17-033-784 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-033-785 Supplier Brady Supplier No. CFB20
Add to Cart
Add to Cart
Enjoy rapid response and none of the labor or mess of sandbags with water-activated Centurion™ Flood Bags.
Transform flood readiness. Maximize asset protection. Enjoy rapid response and none of the labor or mess of sandbags with water-activated Centurion™ Flood Bags. Freshwater-swelled bags contain, control and divert flood water. Absorbent-filled design swells to full size on contact with water in 10 minutes. Flat and lightweight when dry for space-saving storage and easy positioning before first use. Easier and cleaner than sandbags. Multiple bags can be stacked in brick formation to increase barrier height. Cannot be used with salt water.
  • Freshwater-swelled bags contain, control and divert flood water
  • Absorbent-filled design swells to full size on contact with water in 10 minutes
  • Flat and lightweight when dry for space-saving storage and easy positioning before first use
Specifications
Material Polypropylene
Type Spill Control System
Packaging Quantity 200/Cs.
Length (English) 12 in.
Diameter (English) 24 in.