Learn More
Brady Centurion™ Flood Barriers
Catalog No. 17033782 Shop All Brady Corporation Products
Change view
Click to view available options
Length (English):
5 ft.
10 ft.
17 ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
17-033-782 5 ft.
17-033-780 10 ft.
17-033-781 17 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-033-782 Supplier Brady Supplier No. BARRIER5CS
  • $2,755.00
    $429.65 / Case of 10
    Save $2,325.35
    84% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Enjoy rapid response and none of the labor or mess of sandbags with water-activated Centurion™ Flood Barriers.
Transform flood readiness. Maximize asset protection. Enjoy rapid response and none of the labor or mess of sandbags with water-activated Centurion™ Flood Barriers. Freshwater-swelled barriers contain, control and divert flood water. Absorbent-filled design swells to full size on contact with water in 10 minutes. Flat and lightweight when dry for space-saving storage and easy positioning before first use. Easier and cleaner than sandbags. Built-in wedge prevents rolling. To divert flowing water, offset each downstream barrier toward dry side and slightly behind upstream barrier. To stack barriers, place wedge on top of wedge with two barriers, then place a third barrier on top in the middle. Cannot be used with salt water.
  • Freshwater-swelled barriers contain, control and divert flood water
  • Absorbent-filled design swells to full size on contact with water in 10 minutes
  • Flat and lightweight when dry for space-saving storage and easy positioning before first use
Specifications
Material Polypropylene
Type Spill Control System
Packaging Quantity 10/Cs.
Length (English) 5 ft.
Diameter (English) 6 in.