Learn More
Brady Centurion™ Hi-Vis Reinforced Flood Barriers
Catalog No. 17033783 Shop All Brady Corporation Products
Change view
Click to view available options
Type:
Spill Control System
Catalog No. Type
17-033-783 Spill Control System
1 options
Catalog No. 17-033-783 Supplier Brady Supplier No. BARRIERHIVIS
  • $362.00
    $229.65 / Case of 10
    Save $132.35
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Enjoy rapid response and none of the labor or mess of sandbags with water-activated Centurion™ Hi-Vis Reinforced Flood Barriers.
  • Freshwater-swelled barriers contain, control and divert flood water
  • Absorbent-filled design swells to full size on contact with water in 10 minutes
  • Flat and lightweight when dry for space-saving storage and easy positioning before first use
Specifications
Material Polypropylene
Type Spill Control System
Packaging Quantity 10/Pk.
Length (English) 10 ft.
Diameter (English) 6 in.