Learn More
3M™ Centurion™ Safety Goggles
SCI_ED
Catalog No. p-2379234 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Impact-resistant
Impact-resistant
Lens Type:
Chemical-resistant
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Type
S64961 Anti-fog, Impact-resistant Chemical-resistant
S64981 Impact-resistant Chemical-resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S64961 Supplier 3M™ Supplier No. 10078371623903
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Visibility and versatility
  • Wide-angle wraparound lens provides indirect ventilation for chemical splash protection
  • Allows excellent peripheral vision and impact resistance
  • Fits comfortably over most prescription glasses
  • 3M Catalog# 40304-00000-10 , 3M Catalog# 40305-00000-10
Order Info
Available with an optional antifog lens.
Compliance
Meet ANSI Z87.1-2010 and CSA Z94.3 standards
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Impact-resistant
Lens Type Chemical-resistant
Certifications/Compliance ANSI Z87.1-2010 and CSA Z94.3 standards
Lens Tint Clear
Product Type Safety Goggles
Ventilation Details Indirect