Learn More
3M™ Centurion™ Splash Goggles
Catalog No. 19047713 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Uncoated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating
19-047713 Anti-fog
19-130-1362 Uncoated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
Catalog No. 19-047713 Supplier 3M™ Supplier No. 7000002377
Add to Cart
Add to Cart
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
Open viewing range promotes compliance
  • Wide-angle wraparound design offers exceptional side-to-side peripheral vision
  • Polycarbonate lens absorbs 99.9% UV
  • Large lens opening to accommodate prescription and safety eyewear
  • Wide nasal flare
  • Vertical, indirect-venting system to circulate air over lens
  • 3M Catalog# 40304-00000-10 , 3M Catalog# 40305-00000-10
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3-2007 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint Clear
Product Type Splash Goggles
Model 40305-00000-10
Material Polycarbonate