Learn More
Abnova™ CENX CISH Probe
Catalog No. 89335560 Shop All Abnova Corporation Products
Change view
Click to view available options
Product Type:
CENX CISH Probe
Catalog No. Product Type
89-335-560 CENX CISH Probe
1 options
Catalog No. 89-335-560 Supplier Abnova™ Supplier No. CC0002
Add to Cart
Add to Cart
CENX CISH Probe is designed for the qualitative detection of human chromosome X alpha satellites in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH).
Specifications
Content And Storage The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use.
Description The probe is intended to be used in combination with the CISH Implementation Kit 1, which provides necessary reagents for specimen pretreatment and post-hybridization processing.
Product Type CENX CISH Probe
Includes Reagent Provided:
1. Digoxigenin-labeled polynucleotides targeting sequences mapping in Xp11.1-q11.1 specific for the alpha satellite centromeric region DXZ1 of chromosome X
2. Formamide based hybridization buffer