Learn More
Abnova™ CENXp/CENYq CISH Probe
Catalog No. 89335529 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-P
Catalog No. For Use With (Application)
89-335-529 CISH-P
1 options
Catalog No. 89-335-529 Supplier Abnova™ Supplier No. CM0001
Add to Cart
Add to Cart
CENXp/CENYq CISH Probe is designed for the qualitative detection of human chromosome X and chromosome Y alpha satellites in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH).
Reagent Provided:
1. Dinitrophenyl-labeled polynucleotides targeting sequences mapping in Xp11.1-q11.1 specific for the alpha satellite centromeric region DXZ1 of chromosome X.
2. Digoxigenin-labeled polynucleotides targeting sequences mapping in Yp11.1-q11.1 specific for the alpha satellite centromeric region DYZ3 of chromosome Y.
3. Formamide based hybridization buffer.
Specifications
Content And Storage The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use.
Description The probe is intended to be used in combination with the CISH Implementation Kit 2, which provides necessary reagents for specimen pretreatment and post-hybridization processing.
For Use With (Application) CISH-P