Learn More
Apexbio Technology LLC CEP-18770, 10mg. Cas: 847499-27-8 MFCD: MFCD18251439. Proteasome inhibitor
SDP
Catalog No. 501904851
Encompass_Preferred
CEP-18770 is a novel, potent and reversible P2 threonine boronic acid inhibitor of proteasome that inhibits proteasomes chymotrypsin-like activity, with the value of inhibition constant IC50 of 3.8 nM, by down-modulating the activity of nuclear factor-KB (NF-KB). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4851 Supplier Apexbio Technology LLC Supplier No. A400910
CEP-18770 is a novel, potent and reversible P2 threonine boronic acid inhibitor of proteasome that inhibits proteasomes chymotrypsin-like activity, with the value of inhibition constant IC50 of 3.8 nM, by down-modulating the activity of nuclear factor-KB (NF-KB). Other sizes are available. Please inqury us for quote.