Learn More
Apexbio Technology LLC CEP-18770 847499-27-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064195
Encompass_Preferred
CEP-18770 (CAS 847499-27-8) is a reversible boronic acid-based inhibitor of proteasome chymotrypsin-like activity It specifically targets the proteasome s P2 threonine protease subunit reducing NF- B signaling and the expression of downstream effectors With a reported IC50 of 3 8 nM CEP-18770 induces apoptosis in multiple myeloma cell lines and inhibits osteoclastogenesis and angiogenesis via suppression of RANKL signaling Research supports its potential use in oncology particularly for multiple myeloma showing selective cytotoxicity sparing normal epithelial and bone marrow-derived cells
Catalog No. 50-000-64195 Supplier Apexbio Technology LLC Supplier No. A40095.1
 Restricted Item
Add to Cart
Add to Cart
CEP-18770 (CAS 847499-27-8) is a reversible boronic acid-based inhibitor of proteasome chymotrypsin-like activity It specifically targets the proteasome s P2 threonine protease subunit reducing NF- B signaling and the expression of downstream effectors With a reported IC50 of 3 8 nM CEP-18770 induces apoptosis in multiple myeloma cell lines and inhibits osteoclastogenesis and angiogenesis via suppression of RANKL signaling Research supports its potential use in oncology particularly for multiple myeloma showing selective cytotoxicity sparing normal epithelial and bone marrow-derived cells