Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CEP-28122 mesylate salt | 2070009-30-0 | 99.9% | 635.17 | 50 MG
SDP

Catalog No. 5000386869
Encompass_Preferred

CEP-28122 mesylate salt | 2070009-30-0 | 99.9% | 635.17 | 50 MG

Catalog No. 50-003-86869 Supplier Medchemexpress LLC Supplier No. HY18030A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CEP-28122 mesylate salt | 2070009-30-0 | 99.9% | 635.17 | 50 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.