Learn More
TARGETMOL CHEMICALS INC CEP-33779 10MG
SDP
Catalog No. 502579538
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CEP-33779 is a novel and selective inhibitor of JAK2 with an IC50 of 1.8±0.6 nM. purity: 99%
Catalog No. 50-257-9538 Supplier TARGETMOL CHEMICALS INC Supplier No. T612210MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CEP-33779 is a novel and selective inhibitor of JAK2 with an IC50 of 1.8±0.6 nM. purity: 99%