Learn More
Selleck Chemical LLC CEP-33779 5mg 1257704-57-6
SDP
Supplier:  Selleck Chemical LLC S28065MG
Encompass_Preferred
CEP33779 is a selective JAK2 inhibitor with IC50 of 1.8 nM, >40- and >800-fold versus JAK1 and TYK2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8031