Learn More
Medchemexpress LLC Cep-40783 | 1437321-24-8 | 99.6% | 588.56 g/mol | C31H26F2N4O6 | 500 MG
SDP
Catalog No. 5000211645
Encompass_Preferred
CEP-40783 is a potent, selective, orally available small-molecule inhibitor of the receptor tyrosine kinases AXL and c-Met. It exhibits low-nanomolar biochemical and cellular activity and is intended for preclinical research on RTK signaling and oncology models.
  • Potent dual inhibition of AXL and c-Met at low nanomolar concentrations.
  • Demonstrated cellular activity in transfected and tumor cell lines.
  • High purity suitable for biochemical and cellular assays.
  • Powder stable at -20°C for long-term storage; solutions stable at -80°C.
  • Available in a 500 mg package size for research use.
Catalog No. 50-002-11645 Supplier Medchemexpress LLC Supplier No. HY100946500MG
CEP-40783 is a potent, selective, orally available small-molecule inhibitor of the receptor tyrosine kinases AXL and c-Met. It exhibits low-nanomolar biochemical and cellular activity and is intended for preclinical research on RTK signaling and oncology models.
  • Potent dual inhibition of AXL and c-Met at low nanomolar concentrations.
  • Demonstrated cellular activity in transfected and tumor cell lines.
  • High purity suitable for biochemical and cellular assays.
  • Powder stable at -20°C for long-term storage; solutions stable at -80°C.
  • Available in a 500 mg package size for research use.