Learn More
Medchemexpress LLC HY-100946 5mg Medchemexpress, CEP-40783 CAS:1437321-24-8 Purity:>98%
SDP
Catalog No. 501931996
Encompass_Preferred
Medchemexpress, HY-100946 5mg CEP-40783 CAS:1437321-24-8 CEP-40783 is a potent, selective and orally available inhibitor of AXL and c-Met with IC 50 values of 7 nM and 12 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1996 Supplier Medchemexpress LLC Supplier No. HY1009465MG
Medchemexpress, HY-100946 5mg CEP-40783 CAS:1437321-24-8 CEP-40783 is a potent, selective and orally available inhibitor of AXL and c-Met with IC 50 values of 7 nM and 12 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.