Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP 55-Gallon Poly Drum
SDP

Catalog No. 19122696
Encompass
Encompass_Preferred
Change view
Click to view available options
Capacity (English):
55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-122-696 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-122-696 Supplier CEP Supplier No. PD55RS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Durable, chemical-resistant polyethylene drum features:

  • Open head with lever lock ring
  • Incinerable
  • UN rated for salvage drum waste

Specifications

Certifications/Compliance UN
Capacity (English) 55 gal.
Capacity (Metric) 208 L
Color Blue
For Use With (Application) For Salvage Waste
Material Polyethylene
Head Style Open
Includes Lever Lock Ring
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.