Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cep dipeptide 1 | 816432-15-2 | 99.8% | 381.42 | C18H27N3O6 | 10 MG
SDP

Catalog No. 5000295217
Encompass_Preferred

CEP dipeptide 1 is a research-grade CEP-derived dipeptide reported to modulate angiogenic activity relevant to mediators of age-related macular degeneration. It is supplied as a high-purity solid for in vitro research, with stability and solubility data provided for experimental handling.

  • High purity: 99.8%.
  • Molecular weight: 381.42 g/mol.
  • Chemical formula: C18H27N3O6.
  • Soluble in water at about 10 mg/mL; may require ultrasonic treatment.
  • Store powder sealed and away from moisture; -80°C for long-term storage.
  • Available in multiple solid sizes and solution preparations for flexible use.
  • Intended for in vitro research use only; not for human or veterinary applications.

Catalog No. 50-002-95217 Supplier Medchemexpress LLC Supplier No. HY1695910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CEP dipeptide 1 is a research-grade CEP-derived dipeptide reported to modulate angiogenic activity relevant to mediators of age-related macular degeneration. It is supplied as a high-purity solid for in vitro research, with stability and solubility data provided for experimental handling.

  • High purity: 99.8%.
  • Molecular weight: 381.42 g/mol.
  • Chemical formula: C18H27N3O6.
  • Soluble in water at about 10 mg/mL; may require ultrasonic treatment.
  • Store powder sealed and away from moisture; -80°C for long-term storage.
  • Available in multiple solid sizes and solution preparations for flexible use.
  • Intended for in vitro research use only; not for human or veterinary applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.