Learn More
CEP Haz Mat-Aggressive Sorbent Spill Kits
SDP
Catalog No. 19135960
Click to view available options
Capacity (Metric):
18.92 L
Includes:
5 gal. plastic pail with lid, 10 sorbent pads, 2 sorbent socks, 1 pair of goggles, 1 pair of gloves, 1 disposable bag
Catalog No. Capacity (Metric) Includes
19-135-960 18.92 L 5 gal. plastic pail with lid, 10 sorbent pads, 2 sorbent socks, 1 pair of goggles, 1 pair of gloves, 1 disposable bag
1 options
Catalog No. 19-135-960 Supplier CEP Supplier No. HAZSK5
For fast hazardous spill response
Spill kits include sorbents listed in table plus goggles, gloves, and disposable bag within a plastic drum.
Specifications
Capacity (English) 5 gal.
Capacity (Metric) 18.92 L
Type Hazmat Aggressive Sorbent Spill Kit
For Use With (Application) Fast hazardous spill response
Includes 5 gal. plastic pail with lid, 10 sorbent pads, 2 sorbent socks, 1 pair of goggles, 1 pair of gloves, 1 disposable bag